siRNA Details
Gene Symbol: SPRR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006945.4 | NP_008876.3 | 2 | 283 | ||
XM_017002175.1 | XP_016857664.1 | 2 | 315 | |||
GenBank mRNA | AK291317.1 | - | 2 | 285 | ||
BC093801.1 | - | 2 | 267 | |||
BC093803.1 | - | 2 | 268 | |||
BC120938.1 | - | 2 | 267 | 40115065 | ||
BG183546.1 | - | 3 | 512 | |||
M21302.1 | - | 2 | 283 |
Gene Symbol: SPRR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017418.2 | NP_001017418.1 | 2 | 286 | ||
XM_017002174.1 | XP_016857663.1 | 2 | 282 | |||
GenBank mRNA | BF090370.1 | - | 1 | 158 |
Gene Symbol: SPRR2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014450.2 | NP_001014450.1 | 2 | 284 | ||
XM_017002176.1 | XP_016857665.1 | 2 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
