siRNA Details
Gene Symbol: TAF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015975.4 | NP_057059.2 | 7 | 689 | ||
GenBank mRNA | AF220509.1 | - | 7 | 640 | ||
AK312579.1 | - | 7 | 648 | |||
BC009566.1 | - | 7 | 640 | 3996687 | ||
BC010350.1 | - | 7 | 654 | 4276335 | ||
BC071649.1 | - | 7 | 626 | 5246715 | ||
BC146952.1 | - | 7 | 670 | 9056880 | ||
BC146960.1 | - | 7 | 670 | 9056888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
