siRNA Details
Gene Symbol: TAF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015975.4 | NP_057059.2 | 7 | 787 | ||
GenBank mRNA | AF220509.1 | - | 7 | 738 | ||
AK312579.1 | - | 7 | 746 | |||
BC009566.1 | - | 7 | 738 | 3996687 | ||
BC010350.1 | - | 7 | 752 | 4276335 | ||
BC071649.1 | - | 7 | 724 | 5246715 | ||
BC146952.1 | - | 7 | 768 | 9056880 | ||
BC146960.1 | - | 7 | 768 | 9056888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
