siRNA Details
Gene Symbol: ZBTB8OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308135.1 | NP_001295064.1 | 1 | 95 | ||
NM_001308136.1 | NP_001295065.1 | 1 | 95 | |||
NM_001308137.1 | NP_001295066.1 | 1 | 95 | |||
NM_001308138.1 | NP_001295067.1 | 1 | 95 | |||
NM_001308139.1 | NP_001295068.1 | 1 | 95 | |||
NM_001308140.1 | NP_001295069.1 | 1 | 95 | |||
NM_001308141.1 | NP_001295070.1 | 1 | 95 | |||
NM_178547.3 | NP_848642.1 | 1 | 95 | |||
XM_011541327.2 | XP_011539629.1 | 1 | 518 | |||
XM_011541328.1 | XP_011539630.1 | 1 | 50 | |||
XM_011541330.2 | XP_011539632.1 | 1 | 500 | |||
XM_011541334.2 | XP_011539636.1 | 1 | 452 | |||
XM_011541335.2 | XP_011539637.1 | 1 | 30 | |||
XM_011541337.2 | XP_011539639.1 | 1 | 38 | |||
XM_011541338.2 | XP_011539640.1 | 1 | 491 | |||
XM_017001134.1 | XP_016856623.1 | 1 | 40 | |||
XM_017001135.1 | XP_016856624.1 | 1 | 518 | |||
XM_017001136.1 | XP_016856625.1 | 1 | 518 | |||
GenBank mRNA | AA316493.1 | - | 1 | 26 | ||
AK289480.1 | - | 1 | 437 | |||
AY099493.1 | - | 1 | 89 | |||
AY151084.1 | - | 1 | 89 | |||
AY151085.1 | - | 1 | 89 | |||
AY158005.1 | - | 1 | 70 | |||
BC058843.1 | - | 1 | 65 | 6745279 | ||
BF216169.1 | - | 1 | 33 | |||
BG059414.1 | - | 1 | 31 | |||
BM465977.1 | - | 1 | 43 | |||
CN344731.1 | - | 1 | 35 | |||
DT220608.1 | - | 1 | 55 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
