siRNA Details
Gene Symbol: ALYREF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005782.3 | NP_005773.3 | 5 | 711 | ||
GenBank mRNA | AF047002.1 | - | 5 | 612 | ||
AF086513.1 | - | 4 | 366 | |||
BC052302.1 | - | 5 | 711 | 6381528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
