siRNA Details
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020240.1 | - | 2 | 337 | ||
BC041037.1 | - | 2 | 315 | |||
BC073758.1 | - | 2 | 336 | |||
BX640620.1 | - | 2 | 334 | |||
Y14737.1 | - | 2 | 332 |
Gene Symbol: ACSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ496489.1 | - | 210 |
Gene Symbol: IGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX640822.1 | - | 2 | 347 | ||
BX640854.1 | - | 2 | 347 |
Gene Symbol: IGHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF067420.1 | - | 2 | 335 | ||
AK027379.1 | - | 2 | 343 | |||
AK093721.1 | - | 2 | 346 | |||
AK097017.1 | - | 2 | 343 | |||
AK125582.1 | - | 2 | 347 | |||
AK128476.1 | - | 3 | 343 | |||
AK128565.1 | - | 2 | 347 | |||
BC032249.1 | - | 2 | 329 | |||
BC073771.1 | - | 2 | 323 | |||
BX640624.1 | - | 2 | 340 |
Gene Symbol: IGHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128614.1 | - | 2 | 346 | ||
BC021276.2 | - | 2 | 315 |
Gene Symbol: IGHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128313.1 | - | 2 | 346 | ||
AK129508.1 | - | 2 | 347 | |||
BC006402.1 | - | 2 | 341 | |||
BC018747.1 | - | 2 | 329 | |||
BC064496.1 | - | 2 | 335 | |||
BC073782.1 | - | 2 | 329 | |||
BC092518.1 | - | 2 | 327 | |||
BX640627.1 | - | 3 | 347 | |||
Y14735.1 | - | 2 | 302 |
Gene Symbol: IGHG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062335.1 | - | 2 | 330 |
Gene Symbol: IGHV3-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035024.1 | - | 1 | 210 |
Gene Symbol: IGHV3-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035023.1 | - | 2 | 210 |
Gene Symbol: IGHV3-69-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF460695.1 | - | 63 | |||
AF460697.1 | - | 63 | ||||
AF460706.1 | - | 1 | 63 | |||
AF460787.1 | - | 63 | ||||
AF460807.1 | - | 63 | ||||
GU552754.1 | - | 120 | ||||
JX420391.1 | - | 1 | 105 | |||
JX420392.1 | - | 1 | 105 |
Gene Symbol: IGLJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB001733.1 | - | 210 | |||
AB001736.1 | - | 210 |
Gene Symbol: LAIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KU058441.1 | - | 210 |
Gene Symbol: LOC105379528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030112.1 | XP_016885601.1 | 2 | 1389 | ||
GenBank mRNA | Z14178.1 | - | 146 |
Gene Symbol: SCFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Y13056.1 | - | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
