siRNA Details
Gene Symbol: SUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037175.2 | NP_001032252.1 | 4 | 668 | ||
NM_017982.3 | NP_060452.3 | 4 | 668 | |||
XM_005273169.1 | XP_005273226.1 | 5 | 685 | |||
XM_005273172.1 | XP_005273229.1 | 5 | 748 | |||
XM_006711408.1 | XP_006711471.1 | 4 | 666 | |||
XM_011509685.1 | XP_011507987.1 | 4 | 700 | |||
XM_011509687.1 | XP_011507989.1 | 5 | 748 | |||
XM_017001583.1 | XP_016857072.1 | 5 | 866 | |||
XM_017001584.1 | XP_016857073.1 | 5 | 865 | |||
XM_017001585.1 | XP_016857074.1 | 6 | 849 | |||
XM_017001586.1 | XP_016857075.1 | 5 | 743 | |||
XM_017001587.1 | XP_016857076.1 | 5 | 685 | |||
XM_017001588.1 | XP_016857077.1 | 4 | 700 | |||
XM_017001589.1 | XP_016857078.1 | 5 | 870 | |||
GenBank mRNA | AK000914.1 | - | 4 | 549 | ||
AK096265.1 | - | 3 | 1122 | |||
AK295553.1 | - | 3 | 418 | |||
AK296923.1 | - | 5 | 748 | |||
AY358495.1 | - | 4 | 645 | |||
BC004888.2 | - | 4 | 549 | 3926397 | ||
EF560748.1 | - | 4 | 596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
