siRNA Details
Gene Symbol: RFPL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145014.1 | NP_001138486.1 | 3 | 870 | ||
XM_011526915.2 | XP_011525217.1 | 4 | 2158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
