siRNA Details
Gene Symbol: KRBOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129898.1 | NP_001123370.1 | 3, 4 | 322 | ||
NM_001129899.1 | NP_001123371.1 | 3, 4 | 322 | |||
NM_001129900.1 | NP_001123372.1 | 3, 4 | 322 | |||
NM_017776.2 | NP_060246.2 | 3, 4 | 322 | |||
GenBank mRNA | AK000351.1 | - | 3, 4 | 187 | ||
AK096413.1 | - | 3, 4 | 214 | |||
AK289703.1 | - | 3, 4 | 219 | |||
BC012569.1 | - | 2, 3 | 133 | 4283420 | ||
DA152034.1 | - | 3, 4 | 253 | |||
DA631861.1 | - | 3, 4 | 239 | |||
DA877282.1 | - | 3, 4 | 323 |
Gene Symbol: ZNF674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039891.2 | NP_001034980.1 | 3, 4 | 250 | ||
NM_001146291.1 | NP_001139763.1 | 3, 4 | 250 | |||
NM_001190417.1 | NP_001177346.1 | 3, 4 | 250 | |||
XM_011543941.2 | XP_011542243.1 | 3, 4 | 247 | |||
XM_011543943.2 | XP_011542245.1 | 3, 4 | 252 | |||
XM_017029728.1 | XP_016885217.1 | 3, 4 | 213 | |||
XM_017029729.1 | XP_016885218.1 | 3, 4 | 250 | |||
XM_017029730.1 | XP_016885219.1 | 3, 4 | 247 | |||
XM_017029732.1 | XP_016885221.1 | 3, 4 | 213 | |||
XM_017029733.1 | XP_016885222.1 | 3, 4 | 252 | |||
GenBank mRNA | AK295054.1 | - | 3, 4 | 182 | ||
AK308986.1 | - | 3, 4 | 217 | |||
AY971607.1 | - | 3, 4 | 234 | |||
DA195605.1 | - | 3, 4 | 250 | |||
DC317540.1 | - | 3, 4 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
