siRNA Details
Gene Symbol: ZNF57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319083.1 | NP_001306012.1 | 2 | 93 | ||
NM_173480.2 | NP_775751.1 | 2 | 192 | |||
XM_011527682.2 | XP_011525984.1 | 1 | 60 | |||
GenBank mRNA | AK302165.1 | - | 2 | 93 | ||
AK314848.1 | - | 2 | 114 | |||
BC028974.1 | - | 2 | 124 | 5138909 | ||
BX537601.1 | - | 2 | 134 | |||
EF534355.1 | - | 2 | 192 |
Gene Symbol: KRBOX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129898.1 | NP_001123370.1 | 4 | 355 | ||
NM_001129899.1 | NP_001123371.1 | 4 | 355 | |||
NM_001129900.1 | NP_001123372.1 | 4 | 355 | |||
NM_017776.2 | NP_060246.2 | 4 | 355 | |||
GenBank mRNA | AK000351.1 | - | 4 | 220 | ||
AK096413.1 | - | 4 | 247 | |||
AK289703.1 | - | 4 | 252 | |||
BC012569.1 | - | 3 | 166 | 4283420 | ||
DA152034.1 | - | 4 | 286 | |||
DA631861.1 | - | 4 | 272 | |||
DA877282.1 | - | 4 | 356 |
Gene Symbol: ZNF555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172775.1 | NP_001166246.1 | 2 | 182 | ||
NM_152791.4 | NP_690004.4 | 2 | 182 | |||
XM_011527716.2 | XP_011526018.1 | 2 | 182 | |||
XM_017026375.1 | XP_016881864.1 | 2 | 185 | |||
GenBank mRNA | AK093163.1 | - | 2 | 170 | ||
AK292954.1 | - | 2 | 130 | |||
AL832140.1 | - | 2 | 143 | |||
BC022022.2 | - | 2 | 133 | 4822197 | ||
BX647423.1 | - | 2 | 140 |
Gene Symbol: ZNF674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039891.2 | NP_001034980.1 | 4 | 283 | ||
NM_001146291.1 | NP_001139763.1 | 4 | 283 | |||
NM_001190417.1 | NP_001177346.1 | 4 | 283 | |||
XM_011543941.2 | XP_011542243.1 | 4 | 280 | |||
XM_011543943.2 | XP_011542245.1 | 4 | 285 | |||
XM_011543944.2 | XP_011542246.1 | 2 | 90 | |||
XM_017029728.1 | XP_016885217.1 | 4 | 246 | |||
XM_017029729.1 | XP_016885218.1 | 4 | 283 | |||
XM_017029730.1 | XP_016885219.1 | 4 | 280 | |||
XM_017029732.1 | XP_016885221.1 | 4 | 246 | |||
XM_017029733.1 | XP_016885222.1 | 4 | 285 | |||
GenBank mRNA | AK295054.1 | - | 4 | 215 | ||
AK308986.1 | - | 4 | 250 | |||
AY971607.1 | - | 4 | 267 | |||
DA195605.1 | - | 4 | 283 | |||
DC317540.1 | - | 4 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
