siRNA Details
Gene Symbol: DNM1P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003260.1 | 3 | 333 | |||
GenBank mRNA | AK127834.1 | - | 3 | 333 | ||
AK302717.1 | - | 2 | 261 | |||
AL833912.1 | - | 2 | 185 | |||
BC101078.3 | - | 2 | 181 | |||
BC101080.2 | - | 3 | 221 | |||
BC101081.2 | - | 3 | 216 |
Gene Symbol: LOC101927579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720817.3 | XP_006720880.1 | 3 | 211 | ||
XM_011522275.1 | XP_011520577.1 | 5 | 1123 | |||
XM_011522277.1 | XP_011520579.1 | 2 | 436 | |||
XM_011522278.2 | XP_011520580.1 | 5 | 878 | |||
XM_011522279.2 | XP_011520581.1 | 3 | 161 | |||
XM_011522281.2 | XP_011520583.1 | 2 | 143 | |||
XM_011522282.2 | XP_011520584.2 | 7 | 1307 | |||
XM_017022807.1 | XP_016878296.1 | 6 | 703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
