siRNA Details
Gene Symbol: DNM1P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003260.1 | 3 | 308 | |||
GenBank mRNA | AK127834.1 | - | 3 | 308 | ||
AK302717.1 | - | 2 | 236 | |||
AL833912.1 | - | 2 | 160 | |||
BC101078.3 | - | 2 | 156 | |||
BC101080.2 | - | 3 | 196 | |||
BC101081.2 | - | 3 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
