siRNA Details
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027386.2 | 6 | 687 | |||
GenBank mRNA | AK124130.1 | - | 1 | 1855 | ||
BT006760.1 | - | 4 | 362 | |||
DT218991.1 | - | 6 | 608 | |||
X83299.1 | - | 6 | 638 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027028.2 | 6 | 568 | |||
GenBank mRNA | BC147000.1 | - | 6 | 522 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 6 | 709 | |||
GenBank mRNA | BC034017.1 | - | 6 | 710 | ||
BC067351.1 | - | 4 | 407 | |||
BC140728.1 | - | 5 | 532 | |||
BC171739.1 | - | 5 | 532 | 9052954 |
Gene Symbol: GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132999.1 | 4 | 356 | |||
NR_133000.1 | 4 | 356 | ||||
GenBank mRNA | AW955360.1 | - | 3 | 184 |
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 4 | 419 | |||
GenBank mRNA | AW500063.1 | - | 4 | 352 | ||
BC104473.1 | - | 4 | 419 | |||
CF141910.1 | - | 4 | 425 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 6 | 596 | |||
NR_132997.1 | 8 | 855 | ||||
NR_132998.1 | 5 | 523 | ||||
GenBank mRNA | BC144584.1 | - | 6 | 532 | ||
BC144585.1 | - | 5 | 459 | |||
BM553931.1 | - | 8 | 855 | |||
DA269273.1 | - | 5 | 353 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 3 | 313 | |||
NR_029426.1 | 6 | 605 | ||||
GenBank mRNA | AK094021.1 | - | 7 | 1010 | ||
AK097091.1 | - | 7 | 985 | |||
AK289832.1 | - | 6 | 687 | |||
AK315849.1 | - | 6 | 678 | |||
X75940.1 | - | 3 | 314 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 7 | 681 | |||
GenBank mRNA | AK289851.1 | - | 6 | 607 | ||
CN312642.1 | - | 4 | 471 | |||
X83301.1 | - | 7 | 681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
