siRNA Details
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027028.2 | 6 | 545 | |||
GenBank mRNA | AK290693.1 | - | 6 | 512 | ||
BC147000.1 | - | 6 | 499 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 6 | 686 | |||
GenBank mRNA | BC034017.1 | - | 6 | 687 | ||
BC067351.1 | - | 4 | 384 | |||
BC140728.1 | - | 5 | 509 | |||
BC171739.1 | - | 5 | 509 | 9052954 |
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_027386.2 | 6 | 664 | |||
GenBank mRNA | AK124130.1 | - | 1 | 1832 | ||
AK289965.1 | - | 5 | 636 | |||
BT006760.1 | - | 4 | 339 | |||
DT218991.1 | - | 6 | 585 | |||
U21555.1 | - | LIT2100 5G4 pseudogene |
3 | 127 | ||
X83299.1 | - | 6 | 615 |
Gene Symbol: GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132999.1 | 4 | 333 | |||
NR_133000.1 | 4 | 333 | ||||
GenBank mRNA | AW955360.1 | - | 3 | 161 |
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 4 | 396 | |||
GenBank mRNA | AW500063.1 | - | 4 | 329 | ||
BC104473.1 | - | 4 | 396 | |||
CF141910.1 | - | 4 | 402 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 6 | 573 | |||
NR_132997.1 | 8 | 832 | ||||
NR_132998.1 | 5 | 500 | ||||
GenBank mRNA | BC144584.1 | - | 6 | 509 | ||
BC144585.1 | - | 5 | 436 | |||
BM553931.1 | - | 8 | 832 | |||
DA269273.1 | - | 5 | 330 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024054.2 | 3 | 290 | |||
NR_029426.1 | 6 | 582 | ||||
GenBank mRNA | AK094021.1 | - | 7 | 987 | ||
AK097091.1 | - | 7 | 962 | |||
BC131740.1 | - | 4 | 396 | |||
X75940.1 | - | 3 | 291 | |||
X83300.1 | - | 4 | 405 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 7 | 658 | |||
GenBank mRNA | AK289851.1 | - | 6 | 584 | ||
CN312642.1 | - | 4 | 448 | |||
X83301.1 | - | 7 | 658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
