siRNA Details
Gene Symbol: LGALS9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040078.2 | NP_001035167.2 | 10 | 923 | ||
XM_005256769.1 | XP_005256826.1 | 10 | 920 | |||
XM_005256770.1 | XP_005256827.1 | 9 | 824 | |||
XM_011523993.1 | XP_011522295.1 | 9 | 827 | |||
XM_011523994.1 | XP_011522296.1 | 8 | 791 | |||
XM_011523996.1 | XP_011522298.1 | 9 | 827 | |||
XM_011523997.1 | XP_011522299.1 | 8 | 561 | |||
GenBank mRNA | AK315818.1 | - | 10 | 910 | ||
BC073889.1 | - | 10 | 892 | 5222492 | ||
BM769737.1 | - | 6 | 347 |
Gene Symbol: LGALS9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042685.1 | NP_001036150.1 | 10 | 915 | ||
GenBank mRNA | AK290263.1 | - | 10 | 919 | ||
AK296659.1 | - | 9 | 832 | |||
BC105943.1 | - | 10 | 895 | 40031793 | ||
BM769345.1 | - | 6 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
