siRNA Details
Gene Symbol: APOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF305224.1 | - | 1 | 63 |
Gene Symbol: APOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030882.3 | NP_112092.2 | 1 | 178 | ||
NM_145637.2 | NP_663612.2 | 2 | 319 | |||
XM_011530074.1 | XP_011528376.1 | 1 | 117 | |||
XM_011530075.1 | XP_011528377.1 | 2 | 123 | |||
XM_011530076.2 | XP_011528378.1 | 1 | 96 | |||
XM_011530078.1 | XP_011528380.1 | 1 | 117 | |||
XM_017028724.1 | XP_016884213.1 | 1 | 68 | |||
GenBank mRNA | AB208990.1 | - | 1 | 78 | ||
AF305225.1 | - | 1 | 62 | |||
AF305429.1 | - | 1 | 42 | |||
AF324223.1 | - | 1 | 178 | |||
AF324224.1 | - | 2 | 319 | |||
AK056938.1 | - | 1 | 110 | |||
AK303978.1 | - | 1 | 62 | |||
BC004395.1 | - | 1 | 46 | 3636011 |
Gene Symbol: APOL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030643.4 | NP_085146.2 | 1 | 181 | ||
NM_145660.2 | NP_663693.1 | 1 | 181 | |||
GenBank mRNA | AK290466.1 | - | 1 | 62 | ||
AK311506.1 | - | 1 | 154 | |||
AK313637.1 | - | 1 | 118 | |||
AY014914.1 | - | 1 | 182 | |||
AY014915.1 | - | 1 | 181 | |||
BC006276.2 | - | 1 | 91 | |||
DA865632.1 | - | 1 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
