siRNA Details
Gene Symbol: H2AFB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017991.2 | NP_001017991.1 | 1 | 114 | ||
GenBank mRNA | HY000925.1 | - | 1 | 115 |
Gene Symbol: H2AFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017990.1 | NP_001017990.1 | 1 | 41 | ||
GenBank mRNA | BC128034.1 | - | 1 | 33 | 40021540 | |
CR542230.1 | - | 1 | 33 |
Gene Symbol: H2AFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080720.1 | NP_542451.1 | 1 | 41 | ||
GenBank mRNA | AF254576.1 | - | 1 | 41 | ||
BC101409.1 | - | 1 | 33 | 40021547 | ||
BC101415.1 | - | 1 | 33 | 40021541 | ||
BC101417.1 | - | 1 | 33 | 40021543 | ||
BC101418.1 | - | 1 | 33 | 40021544 | ||
BC130510.1 | - | 1 | 109 | 40146541 | ||
BC130512.1 | - | 1 | 109 | 40146543 | ||
BC134365.1 | - | 1 | 36 | 40021542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
