siRNA Details
Gene Symbol: TAF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201536.1 | NP_001188465.1 | 2 | 222 | ||
NM_005681.3 | NP_005672.1 | 2 | 222 | |||
NM_139352.2 | NP_647603.1 | 2 | 222 | |||
XM_005273343.3 | XP_005273400.1 | 2 | 385 | |||
XM_006711612.1 | XP_006711675.1 | 2 | 385 | |||
XM_006711613.3 | XP_006711676.1 | 2 | 217 | |||
XM_017002758.1 | XP_016858247.1 | 2 | 385 | |||
XM_017002759.1 | XP_016858248.1 | 2 | 173 | |||
XM_017002760.1 | XP_016858249.1 | 2 | 169 | |||
GenBank mRNA | AK001054.1 | - | 2 | 74 | ||
AK290970.1 | - | 2 | 167 | |||
AK299536.1 | - | 2 | 222 | |||
AK315740.1 | - | 2 | 206 | |||
BC013808.1 | - | 2 | 202 | 3918866 | ||
L39060.1 | - | 1 | 37 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
