siRNA Details
Gene Symbol: LOC107984026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017029.1 | XP_016872518.1 | 10 | 1739 | ||
XM_017017030.1 | XP_016872519.1 | 9 | 1670 | |||
XM_017017031.1 | XP_016872520.1 | 9 | 1670 | |||
XM_017017032.1 | XP_016872521.1 | 11 | 1770 | |||
XM_017017033.1 | XP_016872522.1 | 12 | 1533 | |||
XM_017017034.1 | XP_016872523.1 | 11 | 1438 | |||
XM_017017035.1 | XP_016872524.1 | 7 | 951 | |||
XM_017017036.1 | XP_016872525.1 | 5 | 1112 | |||
XM_017017037.1 | XP_016872526.1 | 7 | 1501 | |||
XM_017017038.1 | XP_016872527.1 | 5 | 754 | |||
XM_017017039.1 | XP_016872528.1 | 7 | 970 | |||
XM_017017040.1 | XP_016872529.1 | 7 | 934 |
Gene Symbol: AGAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133447.1 | NP_597704.1 | 12 | 1944 | ||
GenBank mRNA | AB075855.1 | - | 7 | 3348 | ||
BC140762.1 | - | 8 | 1190 | 9021630 |
Gene Symbol: AGAP12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029396.2 | 8 | 1238 |
Gene Symbol: AGAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276343.2 | NP_001263272.2 | 8 | 1322 | ||
NM_001291379.1 | NP_001278308.1 | 10 | 1514 | |||
NM_133446.3 | NP_597703.2 | 7 | 1253 | |||
XM_011539269.2 | XP_011537571.1 | 6 | 860 | |||
XM_017015664.1 | XP_016871153.1 | 10 | 5184 | |||
XM_017015665.1 | XP_016871154.1 | 11 | 5052 | |||
XM_017015666.1 | XP_016871155.1 | 9 | 2087 | |||
XM_017015667.1 | XP_016871156.1 | 10 | 1357 | |||
XM_017015668.1 | XP_016871157.1 | 8 | 1107 | |||
XM_017015669.1 | XP_016871158.1 | 8 | 1015 | |||
GenBank mRNA | BC048341.1 | - | 1 | 375 | ||
BC171820.1 | - | 8 | 1307 |
Gene Symbol: AGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144000.1 | NP_001137472.1 | 8 | 1237 |
Gene Symbol: AGAP7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126580.1 | 7 | 1222 | |||
GenBank mRNA | BY996334.1 | - | 1 | 287 |
Gene Symbol: AGAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190810.1 | NP_001177739.1 | 8 | 1308 | ||
GenBank mRNA | AK160375.1 | - | 9 | 4284 | ||
BC171845.1 | - | 8 | 1308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
