siRNA Details
Gene Symbol: IGHV3OR16-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF209733.1 | - | 1 | 170 | ||
AY003842.1 | - | 1 | 162 | |||
DQ454529.1 | - | 1 | 138 | |||
EU099108.1 | - | 1 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
