siRNA Details
Gene Symbol: IGHV3OR16-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF209733.1 | - | 1 | 147 | ||
AY003842.1 | - | 1 | 139 | |||
DQ454529.1 | - | 1 | 115 | |||
EU099108.1 | - | 1 | 154 |
Gene Symbol: IGHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129510.1 | - | 2 | 345 | ||
AK130434.1 | - | 2 | 345 |
Gene Symbol: IGHV4-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY623427.1 | - | 2 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
