siRNA Details
Gene Symbol: PDE3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017912.1 | XP_016873401.1 | 15 | 3286 |
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 61386 |
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008670.1 | - | 2 | 986 |
Gene Symbol: LINC00410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027039.1 | 6 | 692 | |||
GenBank mRNA | AK055781.1 | - | 6 | 692 |
Gene Symbol: LOC101926889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109994.1 | 6 | 1581 | |||
GenBank mRNA | AK092658.1 | - | 6 | 1564 |
Gene Symbol: MIPOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647795.1 | - | 10 | 2984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
