siRNA Details
Gene Symbol: RSPH10B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099697.1 | NP_001093167.1 | 13 | 1744 | ||
XM_005249833.3 | XP_005249890.1 | 7 | 951 | |||
XM_006715766.2 | XP_006715829.1 | 14 | 1896 | |||
XM_006715768.2 | XP_006715831.1 | 7 | 876 | |||
XM_011515491.1 | XP_011513793.1 | 13 | 1788 | |||
XM_011515493.1 | XP_011513795.1 | 13 | 1721 | |||
GenBank mRNA | AK093411.1 | - | 8 | 1081 | ||
AK314983.1 | - | 12 | 1864 | |||
BC044242.1 | - | 9 | 1071 |
Gene Symbol: RSPH10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173565.3 | NP_775836.3 | 13 | 1744 | ||
XM_005249659.3 | XP_005249716.1 | 13 | 2246 | |||
XM_011515203.1 | XP_011513505.1 | 14 | 1841 | |||
XM_011515204.2 | XP_011513506.1 | 13 | 2029 | |||
XM_011515207.1 | XP_011513509.1 | 13 | 1721 | |||
XM_011515210.1 | XP_011513512.1 | 7 | 1003 | |||
XM_011515211.1 | XP_011513513.1 | 7 | 876 | |||
XM_011515212.2 | XP_011513514.1 | 13 | 2261 | |||
XM_017011870.1 | XP_016867359.1 | 13 | 2261 | |||
GenBank mRNA | BC034495.1 | - | 13 | 1747 | 4825868 | |
BC157864.1 | - | 11 | 1494 | 9057083 | ||
BC157871.1 | - | 11 | 1494 | 9057090 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
