siRNA Details
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178087.1 | NP_001171558.1 | 1 | 156 | ||
NM_022978.2 | NP_075267.1 | 1 | 156 | |||
GenBank mRNA | DB454907.1 | - | 1 | 156 |
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 1 | 162 | ||
NM_022968.1 | NP_075257.1 | 1 | 162 | |||
GenBank mRNA | AF073518.1 | - | 1 | 129 | ||
AF073519.1 | - | 1 | 138 | |||
BC021174.1 | - | 1 | 161 | 4824358 | ||
BC035932.1 | - | 1 | 91 | 4701505 | ||
BX107837.1 | - | 1 | 87 | |||
DB444407.1 | - | 1 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
