siRNA Details
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 1 | 197 | ||
NM_022968.1 | NP_075257.1 | 1 | 197 | |||
GenBank mRNA | AF073518.1 | - | 1 | 164 | ||
AF073519.1 | - | 1 | 173 | |||
BC021174.1 | - | 1 | 196 | 4824358 | ||
BC035932.1 | - | 1 | 126 | 4701505 | ||
BX107837.1 | - | 1 | 122 | |||
DB444407.1 | - | 1 | 199 |
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178087.1 | NP_001171558.1 | 1 | 191 | ||
NM_022978.2 | NP_075267.1 | 1 | 191 | |||
GenBank mRNA | BC101970.1 | - | 11 | 40000410 | ||
BC103687.1 | - | 11 | 40000408 | |||
BC104738.1 | - | 11 | 40000411 | |||
BC143260.1 | - | 1 | 13 | 9051762 | ||
BC171749.1 | - | 1 | 13 | 9053250 | ||
DB454907.1 | - | 1 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
