siRNA Details
Gene Symbol: SPATA31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085452.2 | NP_001078921.2 | 4 | 3280 | ||
XM_011518006.1 | XP_011516308.1 | 4 | 3318 | |||
GenBank mRNA | AK302569.1 | - | 4 | 3280 | ||
BC146855.1 | - | 4 | 3239 | 9056783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
