siRNA Details
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 4 | 674 | |||
NR_026982.1 | 3 | 602 | ||||
GenBank mRNA | AK293095.1 | - | 3 | 325 | ||
BC065556.1 | - | 3 | 469 | |||
BQ668440.1 | - | 4 | 674 | |||
DC308368.1 | - | 3 | 394 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 610 |
Gene Symbol: RPL23AP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003572.2 | 4 | 559 | |||
GenBank mRNA | AK127000.1 | - | 3 | 288 | ||
BC052779.1 | - | 5 | 619 | |||
BC064938.1 | - | 4 | 468 |
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 3 | 359 | |||
NR_024528.1 | 4 | 610 | ||||
NR_024529.1 | 3 | 516 | ||||
GenBank mRNA | BC000596.1 | - | 3 | 310 | ||
BC105600.1 | - | 3 | 311 | |||
BG335018.1 | - | 4 | 553 | |||
BX643196.1 | - | 3 | 332 |
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 4 | 1512 | |||
GenBank mRNA | AK125669.1 | - | 4 | 1510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
