siRNA Details
Gene Symbol: RPL23AP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003572.2 | 4 | 607 | |||
GenBank mRNA | AK127000.1 | - | 3 | 336 | ||
BC052779.1 | - | 5 | 667 | |||
BC064938.1 | - | 4 | 516 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 658 |
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 4 | 722 | |||
NR_026982.1 | 3 | 650 | ||||
GenBank mRNA | AK293095.1 | - | 3 | 373 | ||
BC065556.1 | - | 3 | 517 | |||
BQ668440.1 | - | 4 | 722 | |||
DC308368.1 | - | 3 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
