siRNA Details
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 3 | 432 | |||
NR_024528.1 | 4 | 683 | ||||
NR_024529.1 | 3 | 589 | ||||
GenBank mRNA | BC000596.1 | - | 3 | 383 | ||
BC105600.1 | - | 3 | 384 | |||
BG335018.1 | - | 4 | 626 | |||
BX643196.1 | - | 3 | 405 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 683 |
Gene Symbol: RPL23AP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC146939.1 | - | 1 | 311 | 9056867 | |
BC146942.1 | - | 1 | 311 | 9056870 |
Gene Symbol: RPL23AP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003572.2 | 4 | 632 | |||
GenBank mRNA | AK127000.1 | - | 3 | 361 | ||
BC052779.1 | - | 5 | 692 | |||
BC064938.1 | - | 4 | 541 |
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 4 | 747 | |||
NR_026982.1 | 3 | 675 | ||||
GenBank mRNA | AK293095.1 | - | 3 | 398 | ||
BC065556.1 | - | 3 | 542 | |||
BQ668440.1 | - | 4 | 748 | |||
DC308368.1 | - | 3 | 467 |
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 4 | 1585 | |||
GenBank mRNA | AK125669.1 | - | 4 | 1583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
