siRNA Details
Gene Symbol: HLA-F-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026972.1 | 6 | 1602 | |||
NR_026973.1 | 2 | 386 | ||||
GenBank mRNA | AK092748.1 | - | 6 | 1602 | ||
BM726335.1 | - | 2 | 344 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 707 |
Gene Symbol: HLA-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CD369507.1 | - | 1 | 410 |
Gene Symbol: RPL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000984.5 | NP_000975.2 | 3 | 259 | ||
GenBank mRNA | AK130965.1 | - | 3 | 248 | ||
AK312123.1 | - | 3 | 248 | |||
BC014459.1 | - | 1 | 239 | 4872948 | ||
BC058041.1 | - | 3 | 251 | 6716166 | ||
BG476195.1 | - | 3 | 368 | |||
BP382007.1 | - | 3 | 259 | |||
L13799.1 | - | 2 | 209 | |||
U02032.1 | - | 3 | 237 | |||
U43701.1 | - | 3 | 256 |
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 3 | 456 | |||
NR_024528.1 | 4 | 707 | ||||
NR_024529.1 | 3 | 613 | ||||
GenBank mRNA | BC000596.1 | - | 3 | 407 | ||
BC105600.1 | - | 3 | 408 | |||
BX643196.1 | - | 3 | 429 | |||
DA864496.1 | - | 3 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
