siRNA Details
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026981.1 | 4 | 796 | |||
NR_026982.1 | 3 | 724 | ||||
GenBank mRNA | AK293095.1 | - | 3 | 447 | ||
BC065556.1 | - | 3 | 591 | |||
BQ668440.1 | - | 4 | 797 | |||
DC308368.1 | - | 3 | 516 |
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065480.1 | - | 4 | 732 |
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 3 | 481 | |||
NR_024528.1 | 4 | 732 | ||||
NR_024529.1 | 3 | 638 | ||||
GenBank mRNA | BC000596.1 | - | 3 | 432 | ||
BC105600.1 | - | 3 | 433 | |||
BX643196.1 | - | 3 | 454 | |||
DA864496.1 | - | 3 | 488 |
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 4 | 1634 | |||
GenBank mRNA | AK125669.1 | - | 4 | 1632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
