siRNA Details
Gene Symbol: LINC01296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122112.1 | 8 | 1113 | |||
GenBank mRNA | BX248778.1 | - | 8 | 1113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
