siRNA Details
Gene Symbol: LOC107987205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021847.1 | XP_016877336.1 | 1 | 117 |
Gene Symbol: LINC01296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122112.1 | 8 | 1351 | |||
GenBank mRNA | BX248778.1 | - | 8 | 1351 |
Gene Symbol: LOC101929839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546155.1 | XP_011544457.1 | 1 | 117 | ||
GenBank mRNA | AK311726.1 | - | 1 | 117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
