siRNA Details
Gene Symbol: RIMBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015672.1 | NP_056487.1 | 1 | 2991 | ||
GenBank mRNA | AB051453.1 | - | 1 | 2363 | ||
AL133030.1 | - | 1 | 1091 | |||
BC035246.1 | - | 1 | 250 |
Gene Symbol: RIMBP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128635.1 | NP_001122107.1 | 1 | 2614 |
Gene Symbol: RIMBP3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128633.1 | NP_001122105.1 | 1 | 2991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
