siRNA Details
Gene Symbol: RIMBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015672.1 | NP_056487.1 | 1 | 5202 | ||
GenBank mRNA | AB051453.1 | - | 1 | 4574 | ||
AL117509.1 | - | 1 | 658 | |||
AL133030.1 | - | 1 | 3302 | |||
BC035246.1 | - | 1 | 2461 |
Gene Symbol: RIMBP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128635.1 | NP_001122107.1 | 1 | 4825 |
Gene Symbol: RIMBP3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128633.1 | NP_001122105.1 | 1 | 5202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
