siRNA Details
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013453.3 | NP_038481.2 | 1 | 43 | ||
GenBank mRNA | AF098306.1 | - | 1 | 19 | ||
BC069393.1 | - | 1 | 19 | 7262682 | ||
BC069816.1 | - | 1 | 19 | 7262670 | ||
CB115288.1 | - | 1 | 43 |
Gene Symbol: SPANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145662.3 | NP_663695.1 | 1 | 43 | ||
GenBank mRNA | CB110020.1 | - | 1 | 43 | ||
CB113891.1 | - | 1 | 43 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
