siRNA Details
Gene Symbol: ROPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317774.1 | NP_001304703.1 | 5 | 814 | ||
NM_001317775.1 | NP_001304704.1 | 4 | 686 | |||
NM_017578.3 | NP_060048.2 | 6 | 894 | |||
XM_011512933.2 | XP_011511235.1 | 6 | 858 | |||
XM_011512934.2 | XP_011511236.1 | 6 | 969 | |||
GenBank mRNA | AF303889.2 | - | 6 | 894 | ||
BC132744.1 | - | 4 | 650 | 40146766 | ||
BC132746.1 | - | 4 | 651 | 40146768 | ||
BG219480.1 | - | 1 | 162 | |||
BG722939.1 | - | 4 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
