siRNA Details
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 1 | 153 | ||
NM_001278923.1 | NP_001265852.1 | 1 | 153 | |||
NM_001278924.1 | NP_001265853.1 | 1 | 153 | |||
GenBank mRNA | AK313626.1 | - | 1 | 16 | ||
BC008813.2 | - | 1 | 67 | 3958020 | ||
DA698972.1 | - | 1 | 136 |
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040084.2 | NP_001035173.1 | 1 | 116 | ||
NM_001271702.1 | NP_001258631.1 | 1 | 116 | |||
NM_001271703.1 | NP_001258632.1 | 1 | 116 | |||
XM_006717951.3 | XP_006718014.1 | 1 | 70 | |||
GenBank mRNA | AK296976.1 | - | 1 | 20 | ||
AK298761.1 | - | 1 | 96 | |||
BC004376.1 | - | 1 | 13 | 3639064 | ||
BC073755.1 | - | 1 | 47 | 6538697 | ||
CB270161.1 | - | 1 | 102 | |||
DC339657.1 | - | 1 | 116 | |||
Z36830.1 | - | 1 | 27 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
