siRNA Details
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 1 | 206 | ||
NM_001278923.1 | NP_001265852.1 | 1 | 206 | |||
NM_001278924.1 | NP_001265853.1 | 1 | 206 | |||
GenBank mRNA | AK223149.1 | - | 1 | 20 | ||
AK300191.1 | - | 1 | 40 | |||
AK307182.1 | - | 1 | 20 | |||
AK313626.1 | - | 1 | 69 | |||
BC008813.2 | - | 1 | 120 | 3958020 | ||
DA698972.1 | - | 1 | 189 | |||
DC381265.1 | - | 1 | 207 | |||
X16662.1 | - | 1 | 50 |
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040084.2 | NP_001035173.1 | 1 | 169 | ||
NM_001271702.1 | NP_001258631.1 | 1 | 169 | |||
NM_001271703.1 | NP_001258632.1 | 1 | 169 | |||
XM_006717951.3 | XP_006718014.1 | 1 | 123 | |||
XM_011540096.1 | XP_011538398.1 | 2 | 322 | |||
XM_011540098.1 | XP_011538400.1 | 2 | 322 | |||
XM_011540099.1 | XP_011538401.1 | 2 | 322 | |||
XM_011540101.2 | XP_011538403.1 | 2 | 322 | |||
GenBank mRNA | AK296573.1 | - | 1 | 20 | ||
AK296976.1 | - | 1 | 73 | |||
AK298761.1 | - | 1 | 149 | |||
AK300139.1 | - | 1 | 20 | |||
AK300158.1 | - | 1 | 27 | |||
BC004376.1 | - | 1 | 66 | 3639064 | ||
BC073755.1 | - | 1 | 100 | 6538697 | ||
CB270161.1 | - | 1 | 155 | |||
DC339657.1 | - | 1 | 169 | |||
Z36830.1 | - | 1 | 80 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
