siRNA Details
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098845.2 | NP_001092315.2 | 1 | 261 | ||
NM_001278923.1 | NP_001265852.1 | 1 | 261 | |||
NM_001278924.1 | NP_001265853.1 | 1 | 261 | |||
GenBank mRNA | AK223149.1 | - | 1 | 75 | ||
AK300191.1 | - | 1 | 95 | |||
AK307182.1 | - | 1 | 75 | |||
AK313626.1 | - | 1 | 124 | |||
BC008813.2 | - | 1 | 175 | 3958020 | ||
DA698972.1 | - | 1 | 244 | |||
DC381265.1 | - | 1 | 262 | |||
X16662.1 | - | 1 | 105 |
Gene Symbol: ANXA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006717951.3 | XP_006718014.1 | 1 | 178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
