siRNA Details
Gene Symbol: FAM63A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319998.1 | NP_001306927.1 | 2 | 274 | ||
GenBank mRNA | AK125959.1 | - | 2 | 274 |
Gene Symbol: AHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001621.4 | NP_001612.1 | 11 | 4876 | ||
GenBank mRNA | AK130815.1 | - | 1 | 167 |
Gene Symbol: ATP1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303263.1 | NP_001290192.1 | 6 | 1619 | ||
NM_001678.4 | NP_001669.3 | 7 | 2414 | |||
GenBank mRNA | BC126175.1 | - | 7 | 2122 | 8991891 | |
M81181.1 | - | 7 | 2193 | |||
U45945.1 | - | 7 | 1822 |
Gene Symbol: ATP5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK294804.1 | - | 1 | 161 |
Gene Symbol: C21orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM512397.1 | - | 1 | 194 | ||
BU789528.1 | - | 1 | 194 |
Gene Symbol: CAMK2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209474.1 | - | 1 | 3014 |
Gene Symbol: CCNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC026272.1 | - | 5 | 1271 |
Gene Symbol: CNDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131263.1 | - | 2 | 147 |
Gene Symbol: DPPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022821.1 | - | 1 | 574 | ||
AK299290.1 | - | 5 | 1015 |
Gene Symbol: EFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308965.1 | - | 3 | 711 | ||
AK311677.1 | - | 3 | 488 |
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ438905.1 | - | 1 | 1798 |
Gene Symbol: HYKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013619.3 | NP_001013641.2 | 5 | 3433 |
Gene Symbol: LINC00888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038301.1 | 2 | 425 | |||
NR_038302.1 | 2 | 286 | ||||
GenBank mRNA | AK057000.1 | - | 2 | 425 | ||
BX421767.2 | - | 2 | 286 |
Gene Symbol: LOC102724720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010129.1 | XP_016865618.1 | 3 | 1083 |
Gene Symbol: LOC440313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127131.1 | - | 1 | 3570 |
Gene Symbol: LOC441666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024380.1 | 3 | 5280 |
Gene Symbol: LRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122887.1 | - | 1 | 1258 |
Gene Symbol: MFSD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525236.2 | XP_011523538.1 | 13 | 2587 |
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010891.4 | NP_001010891.4 | 8 | 3521 | ||
NM_001167741.1 | NP_001161213.1 | 8 | 3540 | |||
XM_005248495.3 | XP_005248552.1 | 9 | 3612 | |||
XM_017009440.1 | XP_016864929.1 | 7 | 3445 | |||
GenBank mRNA | BC035085.1 | - | 1 | 1775 | ||
BC150295.1 | - | 8 | 3514 | |||
BX538064.1 | - | 8 | 3523 | |||
BX647596.1 | - | 6 | 1847 |
Gene Symbol: PCED1B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026544.1 | 3 | 1569 | |||
GenBank mRNA | AK123977.1 | - | 3 | 1561 |
Gene Symbol: PEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001681.1 | XP_016857170.1 | 3 | 1456 |
Gene Symbol: PLAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518072.2 | XP_011516374.1 | 11 | 3527 | ||
XM_017015319.1 | XP_016870808.1 | 11 | 3610 |
Gene Symbol: PMM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209659.1 | - | 1 | 2397 |
Gene Symbol: PTPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538306.1 | - | 2 | 4078 |
Gene Symbol: RPP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533313.2 | XP_011531615.1 | 6 | 5316 | ||
XM_017005641.1 | XP_016861130.1 | 6 | 5279 |
Gene Symbol: SLC16A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270622.1 | NP_001257551.1 | 6 | 5866 | ||
NM_001270623.1 | NP_001257552.1 | 6 | 5886 | |||
NM_004731.4 | NP_004722.2 | 5 | 5724 | |||
NR_073055.1 | 7 | 6000 | ||||
NR_073056.1 | 7 | 5861 | ||||
XM_005269231.4 | XP_005269288.1 | 5 | 5660 | |||
XM_011538989.2 | XP_011537291.1 | 7 | 6070 | |||
XM_011538990.2 | XP_011537292.1 | 6 | 5962 | |||
XM_011538991.2 | XP_011537293.1 | 6 | 5760 | |||
XM_011538992.2 | XP_011537294.1 | 6 | 6695 | |||
XM_011538993.2 | XP_011537295.1 | 8 | 6460 | |||
XM_011538995.2 | XP_011537297.1 | 8 | 6455 | |||
XM_017020224.1 | XP_016875713.1 | 9 | 6381 | |||
XM_017020225.1 | XP_016875714.1 | 9 | 6374 | |||
XM_017020226.1 | XP_016875715.1 | 9 | 6509 | |||
XM_017020227.1 | XP_016875716.1 | 8 | 6055 |
Gene Symbol: SLC52A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123566.1 | - | 1 | 1089 |
Gene Symbol: SNRNP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK290127.1 | - | 2 | 1254 |
Gene Symbol: SPTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209757.1 | - | 8 | 2023 |
Gene Symbol: TATDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042552.2 | NP_001036017.1 | 10 | 1437 | ||
NM_001042553.2 | NP_001036018.1 | 10 | 1434 | |||
NM_001146169.1 | NP_001139641.1 | 10 | 1424 | |||
NM_001146170.1 | NP_001139642.1 | 9 | 1374 | |||
NM_001146171.1 | NP_001139643.1 | 10 | 1458 | |||
XM_011509202.2 | XP_011507504.1 | 9 | 1319 | |||
XM_017000328.1 | XP_016855817.1 | 9 | 1390 | |||
XM_017000329.1 | XP_016855818.1 | 9 | 1366 | |||
XM_017000330.1 | XP_016855819.1 | 9 | 1351 | |||
XM_017000331.1 | XP_016855820.1 | 9 | 1316 | |||
GenBank mRNA | AK293129.1 | - | 10 | 1458 | ||
AL832248.1 | - | 10 | 1374 | |||
BC048115.1 | - | 10 | 1347 |
Gene Symbol: TMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030755.4 | NP_110382.3 | 8 | 3553 |
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311659.1 | - | 3 | 825 |
Gene Symbol: TRPM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC051024.1 | - | 10 | 2026 |
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094617.1 | - | 1 | 198 |
Gene Symbol: ZMYM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007167.3 | NP_009098.3 | 16 | 4733 | ||
GenBank mRNA | AL833263.1 | - | 15 | 5829 |
Gene Symbol: ZNF316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278559.1 | NP_001265488.1 | 9 | 6104 |
Gene Symbol: ZNF620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017006069.1 | XP_016861558.1 | 1 | 121 |
Gene Symbol: ZNF681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138286.2 | NP_612143.2 | 4 | 3334 |
Gene Symbol: ZSCAN25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160369.1 | - | 1 | 1015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
