siRNA Details
Gene Symbol: KRT39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213656.3 | NP_998821.3 | 3, 4 | 736 | ||
GenBank mRNA | AJ786657.1 | - | 3, 4 | 736 | ||
BC157887.1 | - | 3, 4 | 730 | 8862708 | ||
BC171907.1 | - | 4, 5 | 774 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
