siRNA Details
Gene Symbol: RP9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003500.1 | 3 | 399 | |||
GenBank mRNA | AF274938.1 | - | 2 | 109 | ||
AK091223.1 | - | 2 | 243 | |||
BC050570.1 | - | 4 | 504 |
Gene Symbol: RP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203288.1 | NP_976033.1 | 2 | 185 | ||
XM_011515468.2 | XP_011513770.1 | 2 | 661 | |||
GenBank mRNA | BC025928.1 | - | 2 | 185 | 4903338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
