siRNA Details
Gene Symbol: ZFP64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319146.1 | NP_001306075.1 | 5 | 917 | ||
NM_018197.2 | NP_060667.2 | 6 | 1211 | |||
NM_022088.4 | NP_071371.3 | 5 | 1049 | |||
NM_199426.1 | NP_955458.1 | 6 | 1205 | |||
NM_199427.2 | NP_955459.2 | 9 | 1676 | |||
XM_005260449.1 | XP_005260506.1 | 5 | 628 | |||
XM_006723822.1 | XP_006723885.1 | 5 | 846 | |||
XM_011528900.1 | XP_011527202.1 | 7 | 1102 | |||
XM_017027945.1 | XP_016883434.1 | 9 | 1462 | |||
XM_017027946.1 | XP_016883435.1 | 5 | 771 | |||
XM_017027947.1 | XP_016883436.1 | 8 | 1318 | |||
XM_017027948.1 | XP_016883437.1 | 6 | 750 | |||
XM_017027949.1 | XP_016883438.1 | 7 | 1069 | |||
GenBank mRNA | AK001596.1 | - | 6 | 1001 | ||
AK001744.1 | - | 5 | 776 | |||
AK022690.1 | - | 5 | 917 | |||
AK090547.1 | - | 7 | 873 | |||
BC012759.2 | - | 6 | 1211 | 3634346 | ||
BC021087.1 | - | 9 | 1397 | |||
BC041622.1 | - | 6 | 1211 | 3633938 | ||
BT009760.1 | - | 9 | 1327 |

More Information
Other Species Targeted

Gene Ontology Categories:
