siRNA Details
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 3 | 389 | ||
XM_005255741.3 | XP_005255798.1 | 4 | 1201 | |||
XM_005255742.4 | XP_005255799.1 | 4 | 947 | |||
XM_011545929.2 | XP_011544231.1 | 5 | 1021 | |||
XM_011545931.2 | XP_011544233.1 | 4 | 654 | |||
XM_017023628.1 | XP_016879117.1 | 4 | 404 | |||
XM_017023629.1 | XP_016879118.1 | 4 | 947 | |||
XM_017023630.1 | XP_016879119.1 | 5 | 674 | |||
XM_017023631.1 | XP_016879120.1 | 3 | 1018 | |||
XM_017023632.1 | XP_016879121.1 | 4 | 404 | |||
GenBank mRNA | BC144439.1 | - | 3 | 350 | ||
BC157836.1 | - | 3 | 1039 | 9057055 | ||
KF175222.1 | - | 3 | 351 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 3 | 351 | ||
XM_017023622.1 | XP_016879111.1 | 4 | 475 | |||
XM_017023623.1 | XP_016879112.1 | 4 | 475 | |||
XM_017023624.1 | XP_016879113.1 | 4 | 475 | |||
XM_017023625.1 | XP_016879114.1 | 4 | 455 | |||
XM_017023626.1 | XP_016879115.1 | 5 | 1094 | |||
XM_017023627.1 | XP_016879116.1 | 4 | 475 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 3 | 389 | ||
NM_001287251.1 | NP_001274180.1 | 4 | 475 | |||
XM_003118719.7 | XP_003118767.2 | 4 | 950 | |||
XM_011545690.2 | XP_011543992.1 | 5 | 1039 | |||
XM_011545693.2 | XP_011543995.1 | 5 | 1038 | |||
XM_017022814.1 | XP_016878303.1 | 5 | 1039 | |||
XM_017022815.1 | XP_016878304.1 | 5 | 559 | |||
XM_017022816.1 | XP_016878305.1 | 5 | 1039 | |||
XM_017022817.1 | XP_016878306.1 | 4 | 951 | |||
XM_017022818.1 | XP_016878307.1 | 4 | 2276 | |||
XM_017022819.1 | XP_016878308.1 | 5 | 966 | |||
XM_017022820.1 | XP_016878309.1 | 5 | 683 | |||
XM_017022821.1 | XP_016878310.1 | 4 | 1549 | |||
XM_017022822.1 | XP_016878311.1 | 5 | 2552 | |||
XM_017022823.1 | XP_016878312.1 | 3 | 2188 | |||
XM_017022824.1 | XP_016878313.1 | 5 | 965 | |||
XM_017022825.1 | XP_016878314.1 | 5 | 683 | |||
XM_017022826.1 | XP_016878315.1 | 4 | 500 | |||
XM_017022828.1 | XP_016878317.1 | 5 | 1038 | |||
GenBank mRNA | BC171825.1 | - | 4 | 1125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
