siRNA Details
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022847.1 | XP_016878336.1 | 7 | 716 | ||
GenBank mRNA | BY996642.1 | - | 3 | 284 |
Gene Symbol: LOC105376752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522797.2 | XP_011521099.1 | 6 | 726 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK294177.1 | - | 3 | 180 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522587.1 | XP_011520889.1 | 9 | 3166 | ||
XM_011522596.1 | XP_011520898.1 | 6 | 622 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522585.1 | XP_011520887.1 | 6 | 622 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522329.2 | XP_011520631.1 | 9 | 1114 | ||
XM_011522337.1 | XP_011520639.1 | 6 | 622 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022840.1 | XP_016878329.1 | 7 | 716 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308629.1 | - | 1 | 431 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023622.1 | XP_016879111.1 | 7 | 881 | ||
XM_017023623.1 | XP_016879112.1 | 7 | 881 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022814.1 | XP_016878303.1 | 8 | 1445 | ||
XM_017022815.1 | XP_016878304.1 | 8 | 965 | |||
XM_017022816.1 | XP_016878305.1 | 8 | 1445 | |||
XM_017022817.1 | XP_016878306.1 | 7 | 1357 | |||
XM_017022818.1 | XP_016878307.1 | 7 | 2682 | |||
XM_017022819.1 | XP_016878308.1 | 8 | 1372 | |||
XM_017022820.1 | XP_016878309.1 | 8 | 1089 | |||
XM_017022821.1 | XP_016878310.1 | 7 | 1955 | |||
XM_017022822.1 | XP_016878311.1 | 8 | 2958 | |||
XM_017022823.1 | XP_016878312.1 | 6 | 2594 | |||
XM_017022826.1 | XP_016878315.1 | 7 | 906 | |||
XM_017022827.1 | XP_016878316.1 | 3 | 653 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC144350.1 | - | 6 | 652 | 9052870 | |
BX647358.1 | - | 2 | 4027 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298381.1 | - | 5 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
