siRNA Details
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 1 | 56 | ||
XM_011523498.2 | XP_011521800.1 | 2 | 146 | |||
XM_017023248.1 | XP_016878737.1 | 2 | 549 | |||
XM_017023249.1 | XP_016878738.1 | 2 | 549 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255741.3 | XP_005255798.1 | 2 | 938 | ||
XM_017023630.1 | XP_016879119.1 | 3 | 411 | |||
XM_017023631.1 | XP_016879120.1 | 2 | 938 | |||
GenBank mRNA | BC144439.1 | - | 1 | 87 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023626.1 | XP_016879115.1 | 3 | 831 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022820.1 | XP_016878309.1 | 3 | 420 | ||
XM_017022821.1 | XP_016878310.1 | 2 | 1286 | |||
XM_017022822.1 | XP_016878311.1 | 3 | 2289 | |||
XM_017022825.1 | XP_016878314.1 | 3 | 420 | |||
XM_017022826.1 | XP_016878315.1 | 3 | 420 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC144350.1 | - | 1 | 88 | 9052870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
