siRNA Details
Gene Symbol: MIPOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647795.1 | - | 10 | 5152 |
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 6341 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 5407 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 6208 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 5895 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 6470 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 6734 | |||
NM_003879.5 | NP_003870.4 | 10 | 6313 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 7208 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 6177 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 6256 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 7143 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 9160 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 7016 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 6354 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 5979 |
Gene Symbol: KCNQ1OT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002728.3 | 1 | 63554 |
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 1688 | |||
GenBank mRNA | BC036758.2 | - | 7 | 1700 | 5563071 |
Gene Symbol: LOC100131257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034022.1 | 1 | 11094 |
Gene Symbol: SMC5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR936772.1 | - | 3 | 5350 |
Gene Symbol: TBC1D8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648230.1 | - | 7 | 2709 |
Gene Symbol: UGDH-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047679.1 | 5 | 1566 | |||
GenBank mRNA | AK056558.1 | - | 5 | 1386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
