siRNA Details
Gene Symbol: RPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029.3 | NP_001020.2 | 2, 3 | 453 | ||
GenBank mRNA | AV681946.1 | - | 2, 3 | 453 | ||
BC002604.1 | - | 2, 3 | 417 | 3143099 | ||
BC013215.2 | - | 1 | 163 | |||
BC015832.1 | - | 1 | 214 | 4807004 | ||
BC070220.1 | - | 1 | 214 | 6388128 | ||
BC105276.1 | - | 1 | 217 | 4287636 | ||
BC105798.1 | - | 1 | 218 | 6726218 | ||
X69654.1 | - | 1 | 211 | |||
X77770.1 | - | 1 | 188 |
Gene Symbol: RPS26P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002309.1 | 1 | 324 | |||
GenBank mRNA | AW972305.1 | - | 1 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
