siRNA Details
Gene Symbol: UAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207309.2 | NP_997192.2 | 9 | 3137 | ||
GenBank mRNA | AK022632.1 | - | 4 | 2022 | ||
AK074143.1 | - | 4 | 4186 | |||
AK095393.1 | - | 9 | 2878 | |||
AK128529.1 | - | 7 | 2972 | |||
AL832421.1 | - | 9 | 2906 | |||
BC106035.1 | - | 9 | 2851 |
Gene Symbol: CNBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304367.1 | NP_001291296.1 | 2 | 1150 | ||
GenBank mRNA | BC030782.1 | - | 2 | 1125 | ||
BC057288.1 | - | 2 | 1120 |
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 15 | 10173 | ||
NM_018212.4 | NP_060682.2 | 14 | 10110 | |||
XM_005273187.2 | XP_005273244.1 | 16 | 10213 | |||
XM_017001746.1 | XP_016857235.1 | 17 | 11448 | |||
XM_017001751.1 | XP_016857240.1 | 14 | 10458 | |||
XM_017001753.1 | XP_016857242.1 | 18 | 10252 | |||
XM_017001754.1 | XP_016857243.1 | 17 | 10196 | |||
XM_017001755.1 | XP_016857244.1 | 17 | 9939 | |||
XM_017001756.1 | XP_016857245.1 | 15 | 9870 | |||
XM_017001757.1 | XP_016857246.1 | 15 | 9773 | |||
GenBank mRNA | BC037319.1 | - | 1 | 247 |
Gene Symbol: S1PR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166215.1 | NP_001159687.1 | 2 | 2407 | ||
NM_030760.4 | NP_110387.1 | 2 | 2291 | |||
GenBank mRNA | AF331840.1 | - | 1 | 2236 |
Gene Symbol: SCLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127520.1 | - | 1 | 640 |
Gene Symbol: SERINC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006811.3 | NP_006802.1 | 10 | 3425 | ||
XM_017027601.1 | XP_016883090.1 | 10 | 3319 |
Gene Symbol: TMEM134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545268.2 | XP_011543570.1 | 5 | 1690 | ||
GenBank mRNA | AY007143.1 | - | 4 | 1812 |
Gene Symbol: TMEM59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129688.1 | - | 1 | 879 |
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130467.1 | - | 1 | 1280 |
Gene Symbol: TSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB190910.1 | - | 12 | 1847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
